missing translation for 'onlineSavingsMsg'
Learn More

[N-(2-Maleimidoethyl]ethylenediamine-N,N,N’,N’-tetraacetic Acid, Monoamide, Technical Grade, TRC

Product Code. 30477663
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477663 50 mg 50mg
30475066 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477663 Supplier Toronto Research Chemicals Supplier No. TRCM13848050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185102-85-5
Molecular Formula C16H22N4O9
Molecular Weight (g/mol) 414.37
Synonym [N-(2-Maleimidoethyl]Ethylenediamine-N,N,N',N'-Tetraacetic Acid, Monoamide
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-[2-[2-(2,5-dioxopyrrolidin-1-yl)ethylamino]-2-oxoethyl]amino]acetic acid
SMILES OC(=O)CN(CCN(CC(=O)O)CC(=O)NCCN1C(=O)CCC1=O)CC(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H24N4O9/c21-11(17-3-4-20-12(22)1-2-13(20)23)7-18(8-14(24)25)5-6-19(9-15(26)27)10-16(28)29/h1-10H2,(H,17,21)(H,24,25)(H,26,27)(H,28,29)
Formula Weight 416.1543
Chemical Name or Material [N-(2-Maleimido)ethyl]ethylenediamine-N,N,N',N'-tetraacetic Acid, Monoamide, Technical Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.