missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(4-Aminophenyl)propyl]-N,N-dimethylamine, TRC

Product Code. 30323666
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323666 100 mg 100mg
30327225 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30323666 Supplier Toronto Research Chemicals Supplier No. TRCA615935100MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-(4-Aminophenyl)propyl]-N,N-dimethylamine
CAS 42817-60-7
Molecular Formula C11H18N2
Quantity 100 mg
Synonym 4-Amino-N,N-dimethylbenzenepropanamine, 4-(3-Dimethylaminopropyl)aniline, 4-(3-Dimethylaminopropyl)phenylamine, p-[3-(Dimethylamino)propyl]aniline
InChI Formula InChI=1S/C11H18N2/c1-13(2)9-3-4-10-5-7-11(12)8-6-10/h5-8H,3-4,9,12H2,1-2H3
SMILES CN(C)CCCc1ccc(N)cc1
IUPAC Name 4-[3-(dimethylamino)propyl]aniline
Molecular Weight (g/mol) 178.27
Formula Weight 178.147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.