missing translation for 'onlineSavingsMsg'
Learn More

N-[3-[[4-(Chloromethyl)benzoyl]amino][1,1'-biphenyl]-4-yl]carbamic Acid tert-Butyl Ester, TRC

Product Code. 30367305
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367305 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367305 Supplier Toronto Research Chemicals Supplier No. TRCC36967050MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3-((4-(Chloromethyl)benzoyl)amino)(1,1'-biphenyl)-4-yl)carbamic Acid tert-Butyl Ester
CAS 1003316-10-6
Molecular Formula C25H25ClN2O3
Quantity 50 mg
Synonym N-[3-[[4-(Chloromethyl)benzoyl]amino][1,1'-biphenyl]-4-yl]carbamic Acid 1,1-Dimethylethyl Ester
InChI Formula InChI=1S/C25H25ClN2O3/c1-25(2,3)31-24(30)28-21-14-13-20(18-7-5-4-6-8-18)15-22(21)27-23(29)19-11-9-17(16-26)10-12-19/h4-15H,16H2,1-3H3,(H,27,29)(H,28,30)
SMILES ClCC1=CC=C(C(NC2=CC(C3=CC=CC=C3)=CC=C2NC(OC(C)(C)C)=O)=O)C=C1
IUPAC Name tert-butyl (3-(4-(chloromethyl)benzamido)-[1,1'-biphenyl]-4-yl)carbamate
Molecular Weight (g/mol) 436.93
Formula Weight 436.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.