missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-methoxyphenyl)-2-fluorobenzamide, 97%

Brand:  Alfa Aesar™ H59285.03

Code : NEW

Additional Details : CAS Number : 415943-80-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15474926

  • 646.00 € / 1g

Estimated Shipment date
from Supplier 18-07-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

415943-80-5
279.695
PXQDULFVCYBPCL-UHFFFAOYSA-N
742747
COC1=C(C=C(C=C1)NC(=O)C2=CC=CC=C2F)Cl
C14H11ClFNO2
MFCD00815981
n-3-chloro-4-methoxyphenyl-2-fluorobenzamide, cambridge id 5403321
N-(3-chloro-4-methoxyphenyl)-2-fluorobenzamide

Specifications

N-(3-Chloro-4-methoxyphenyl)-2-fluorobenzamide
415943-80-5
MFCD00815981
PXQDULFVCYBPCL-UHFFFAOYSA-N
N-(3-chloro-4-methoxyphenyl)-2-fluorobenzamide
742747
Solid
1g
C14H11ClFNO2
n-3-chloro-4-methoxyphenyl-2-fluorobenzamide, cambridge id 5403321
COC1=C(C=C(C=C1)NC(=O)C2=CC=CC=C2F)Cl
279.695
279.7
97%
Videos
SDS