missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(Dimethylamino)propyl] Perfluorohexanesulfonamide (>85%), TRC

Artikelnummer. 30138121
Ansicht ändern
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Packungsgröße:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
30138121 250 mg 250mg
30136827 100 mg 100mg
17912675 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30138121 Lieferant Toronto Research Chemicals Supplier No. TRCD616080250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50598-28-2
Molecular Formula C11 H13 F13 N2 O2 S
Molecular Weight (g/mol) 484.2774
Synonym 1-Hexanesulfonamide, N-[3-(dimethylamino)propyl]-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluoro-, N-[3-(Dimethylamino)propyl]-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluoro-1-hexanesulfonamide, N-[3-(Dimethylamino)propyl]perfluoro-1-hexanesulfonamide
IUPAC Name N-[3-(dimethylamino)propyl]-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane-1-sulfonamide
SMILES CN(C)CCCNS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H13F13N2O2S/c1-26(2)5-3-4-25-29(27,28)11(23,24)9(18,19)7(14,15)6(12,13)8(16,17)10(20,21)22/h25H,3-5H2,1-2H3
Formula Weight 484.049
Chemical Name or Material N-[3-(Dimethylamino)propyl]perfluoro-1-hexanesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.