missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Hydroxyphenyl)maleimide, TRC

Product Code. 30392962
Change view
Click to view available options
Quantity:
25 mg
50 mg
200 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392962 200 mg
30406240 50 mg
30406087 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392962 Supplier Toronto Research Chemicals Supplier No. TRCH587090200MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63381-38-4
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
Synonym 1H-Pyrrole-2,5-dione, 1-(3-hydroxyphenyl)- (9CI, ACI), 1-(3-Hydroxyphenyl)-1H-pyrrole-2,5-dione (ACI), 1-(3-Hydroxyphenyl)-2,5-dihydro-1H-pyrrole-2,5-dione, m-Maleimidophenol, N-(3-Hydroxyphenyl)maleimide, N-(m-Hydroxyphenyl)maleimide
IUPAC Name 1-(3-hydroxyphenyl)pyrrole-2,5-dione
SMILES Oc1cccc(c1)N2C(=O)C=CC2=O

Specifications

Quantity 200 mg
InChI Formula InChI=1S/C10H7NO3/c12-8-3-1-2-7(6-8)11-9(13)4-5-10(11)14/h1-6,12H
Formula Weight 189.0426
Chemical Name or Material N-(3-Hydroxyphenyl)maleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.