missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Hydroxypropyl)phthalimide, 98%

Product Code. 11321617
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321617 10 g 10g
11331617 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11321617 Supplier Thermo Scientific Alfa Aesar Supplier No. L04507.09
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 883-44-3 | C11H11NO3 | 205.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 883-44-3
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD00023097
InChI Key BSMILTTURCQDGJ-UHFFFAOYSA-N
Synonym n-3-hydroxypropyl phthalimide, 2-3-hydroxypropyl isoindoline-1,3-dione, n-3-hydroxypropyl-phthalimide, 1h-isoindole-1,3 2h-dione, 2-3-hydroxypropyl, 3-phthalimido-1-propanol, 2-3-hydroxypropyl isoindole-1,3-dione, 2-3-hydroxypropyl-1h-isoindole-1,3 2h-dione, 3-phthalimidopropanol, 3-phthalimidylpropanol, acmc-209qsg
PubChem CID 70160
IUPAC Name 2-(3-hydroxypropyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCO

Specifications

Melting Point 75°C to 80°C
Quantity 10 g
Beilstein 158489
Formula Weight 205.22
Percent Purity 98%
Chemical Name or Material N-(3-Hydroxypropyl)phthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.