missing translation for 'onlineSavingsMsg'
Learn More

N-(3-hydroxypropyl)phthalimide, 99%

Product Code. 10736183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10736183 25 g Glass bottle 25g
10441543 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10736183 Supplier Thermo Scientific Acros Supplier No. 377010250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 883-44-3 | C11H11NO3 | 205.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 883-44-3
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
MDL Number MFCD00023097
InChI Key BSMILTTURCQDGJ-UHFFFAOYSA-N
Synonym n-3-hydroxypropyl phthalimide, 2-3-hydroxypropyl isoindoline-1,3-dione, n-3-hydroxypropyl-phthalimide, 1h-isoindole-1,3 2h-dione, 2-3-hydroxypropyl, 3-phthalimido-1-propanol, 2-3-hydroxypropyl isoindole-1,3-dione, 2-3-hydroxypropyl-1h-isoindole-1,3 2h-dione, 3-phthalimidopropanol, 3-phthalimidylpropanol, acmc-209qsg
PubChem CID 70160
IUPAC Name 2-(3-hydroxypropyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 75°C to 77°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: < 1g/l
Formula Weight 205.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-(3-Hydroxypropyl)phthalimide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.