missing translation for 'onlineSavingsMsg'
Learn More

N-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl Lansoprazole, TRC

Product Code. 30470538
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470538 100 mg 100mg
30466037 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30470538 Supplier Toronto Research Chemicals Supplier No. TRCM331010100MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1083100-26-8
Molecular Formula C25H22F6N4O3S
Molecular Weight (g/mol) 572.52
Synonym 1-[[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]-2-[[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]sulfinyl]-1H-benzimidazole, Lansoprazole Impurity
IUPAC Name 1-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methyl]-2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]benzimidazole
SMILES Cc1c(OCC(F)(F)F)ccnc1CS(=O)c2nc3ccccc3n2Cc4nccc(OCC(F)(F)F)c4C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C25H22F6N4O3S/c1-15-18(32-9-7-21(15)37-13-24(26,27)28)11-35-20-6-4-3-5-17(20)34-23(35)39(36)12-19-16(2)22(8-10-33-19)38-14-25(29,30)31/h3-10H,11-14H2,1-2H3
Formula Weight 572.1317
Chemical Name or Material N-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl Lansoprazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.