missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Pyridyl)thiourea, 98+%

Product Code. 11324534
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324534 5 g 5g
11314534 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324534 Supplier Thermo Scientific Alfa Aesar Supplier No. L01284.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 30162-37-9 | C6H7N3S | 153.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30162-37-9
Molecular Formula C6H7N3S
Molecular Weight (g/mol) 153.20
MDL Number MFCD00014635
InChI Key CFOJQUGXHMGMOT-UHFFFAOYSA-N
Synonym 3-pyridylthiourea, 1-3-pyridyl-2-thiourea, 1-pyridin-3-yl thiourea, pyridin-3-yl-thiourea, n-3-pyridyl thiourea, n-pyridin-3-ylthiourea, pyridin-3-yl thiourea, 1-pyridin-3-ylthiourea, amino 3-pyridylamino methane-1-thione, 3-pyridinylthiourea
PubChem CID 2760515
IUPAC Name pyridin-3-ylthiourea
SMILES NC(=S)NC1=CC=CN=C1

Specifications

Melting Point 158°C to 162°C
Quantity 5 g
UN Number UN2811
Beilstein 472146
Formula Weight 153.21
Percent Purity ≥98%
Chemical Name or Material N-(3-Pyridyl)thiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.