Learn More
LNI Swissgas N.3 Refill for Desiccant Column
Product Code. 17852029 Shop All LNI Swissgas Products
Change view
Click to view available options
Product Type:
Desiccant Column Refill
Unit Size:
Each
Product Code. Product Type Unit Size
17852029 Desiccant Column Refill Each
1 options
Product Code. 17852029 Supplier LNI Swissgas Supplier No. 6930.00.093
  • ONLINE EXCLUSIVE PRICE
    149.00 €
    128.46 € / Pack of 1
    Save 20.54 €
    13% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
N.3 refill for desiccant column.
  • For hydrogen range
  • Replace every 6 months
Specifications
Product Type Desiccant Column Refill