missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromobutyl)phthalimide, 96%

Product Code. 11461868
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461868 100 g 100g
11441868 5 g 5g
11451868 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11461868 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur A14517.22
 Restricted Item
Estimated Shipment: 15-10-2026
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 5394-18-3 | C12H12BrNO2 | 282.14 g/mol

N-(4-Bromobutyl)phthalimide is used in organic synthesis and the production of pharmaceutical. It can react with 1-phenyl-piperazine to get N-[4-(4-phenyl-piperazin-1-yl)-butyl]-phthalimide. It is a useful synthesis reagent used to synthesize B-cyclodextrin derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(4-Bromobutyl)phthalimide is used in organic synthesis and the production of pharmaceutical. It can react with 1-phenyl-piperazine to get N-[4-(4-phenyl-piperazin-1-yl)-butyl]-phthalimide. It is a useful synthesis reagent used to synthesize B-cyclodextrin derivatives.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store at room temperature. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 5394-18-3
Molecular Formula C12H12BrNO2
Molecular Weight (g/mol) 282.14
MDL Number MFCD00005905
InChI Key UXFWTIGUWHJKDD-UHFFFAOYSA-N
Synonym n-4-bromobutyl phthalimide, 2-4-bromobutyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-bromobutyl, 2-4-bromobutyl-1h-isoindole-1,3 2h-dione, n-4-bromobutyl-phthalimide, 2-4-bromobutyl isoindole-1,3-dione, 2-4-bromobutyl-2,3-dihydro-1h-isoindole-1,3-dione, n-4-bromobutyl, n-bromobutylphthalimide
PubChem CID 93575
IUPAC Name 2-(4-bromobutyl)isoindole-1,3-dione
SMILES BrCCCCN1C(=O)C2=CC=CC=C2C1=O

Spécification

Melting Point 77°C to 81°C
Boiling Point 165°C to 170°C (1 mmHg)
Odor Odorless
Quantity 100 g
Beilstein 164119
Solubility Information Soluble in ethanol. Insoluble in water.
Formula Weight 282.14
Percent Purity 98%
Chemical Name or Material N-(4-Bromobutyl)phthalimide

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.