missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromobutyl)phthalimide, 98%

Product Code. 10571961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10571961 10 g Glass bottle 10g
10023663 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10571961 Supplier Thermo Scientific Acros Supplier No. 146290100
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5394-18-3 | C12H12BrNO2 | 282.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5394-18-3
Molecular Formula C12H12BrNO2
Molecular Weight (g/mol) 282.14
MDL Number MFCD00005905
InChI Key UXFWTIGUWHJKDD-UHFFFAOYSA-N
Synonym n-4-bromobutyl phthalimide, 2-4-bromobutyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-bromobutyl, 2-4-bromobutyl-1h-isoindole-1,3 2h-dione, n-4-bromobutyl-phthalimide, 2-4-bromobutyl isoindole-1,3-dione, 2-4-bromobutyl-2,3-dihydro-1h-isoindole-1,3-dione, n-4-bromobutyl, n-bromobutylphthalimide
PubChem CID 93575
IUPAC Name 2-(4-bromobutyl)isoindole-1,3-dione
SMILES BrCCCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 76.0°C to 80.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 21, 463
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 282.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-(4-Bromobutyl)phthalimide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.