missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Cyanobutyl)-N-(phenylmethoxy)-carbamic Acid 1,1-Dimethylethyl Ester, TRC

Product Code. 30353905
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353905 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353905 Supplier Toronto Research Chemicals Supplier No. TRCC95575525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(4-Cyanobutyl)-N-(phenylmethoxy)-carbamic Acid 1,1-Dimethylethyl Ester
CAS 128173-50-2
Molecular Formula C17H24N2O3
Quantity 25 mg
Synonym (4-Cyanobutyl)(phenylmethoxy)-carbamic Acid 1,1-Dimethylethyl Ester, t-Butyl Benzyloxy(4-cyanobutyl)carbamate
InChI Formula InChI=1S/C17H24N2O3/c1-17(2,3)22-16(20)19(13-9-5-8-12-18)21-14-15-10-6-4-7-11-15/h4,6-7,10-11H,5,8-9,13-14H2,1-3H3
SMILES N#CCCCCN(OCC1=CC=CC=C1)C(OC(C)(C)C)=O
IUPAC Name tert-butyl (benzyloxy)(4-cyanobutyl)carbamate
Molecular Weight (g/mol) 304.38
Formula Weight 304.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.