missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Ethoxyphenyl)maleamic acid, 97%, Thermo Scientific™

Product Code. 11425987
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425987 5 g 5g
11435987 25 g 25g
11445987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425987 Supplier Thermo Scientific Alfa Aesar Supplier No. B25024.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108087-84-9
Molecular Formula C12H13NO4
Molecular Weight (g/mol) 235.239
MDL Number MFCD00020477
InChI Key LMTIUWRMRAZPEN-FPLPWBNLSA-N
Synonym z-4-4-ethoxyphenyl amino-4-oxobut-2-enoic acid, 2z-4-4-ethoxyphenyl amino-4-oxobut-2-enoic acid, n-4-ethoxyphenyl maleamic acid, 2z-3-n-4-ethoxyphenyl carbamoyl prop-2-enoic acid, 2z-3-4-ethoxyphenyl carbamoyl prop-2-enoic acid, 4-4-ethoxyanilino-4-oxobut-2-enoic acid, z-4-4-ethoxyanilino-4-oxobut-2-enoic acid, 2-butenoic acid,4-4-ethoxyphenyl amino-4-oxo-, 2z
PubChem CID 1808883
IUPAC Name (Z)-4-(4-ethoxyanilino)-4-oxobut-2-enoic acid
SMILES CCOC1=CC=C(C=C1)NC(=O)C=CC(=O)O

Specifications

Melting Point 188°C to 189°C
Quantity 5 g
Formula Weight 235.24
Percent Purity 97%
Chemical Name or Material N-(4-Ethoxyphenyl)maleamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.