missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]-acetamide, TRC

Product Code. 30478551
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478551 10 mg 10mg
30476982 25 mg 25mg
30473544 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478551 Supplier Toronto Research Chemicals Supplier No. TRCM33024010MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 893990-34-6
Molecular Formula C16H14N4OS2
Molecular Weight (g/mol) 342.44
Synonym CID-7211972, VU0240551
IUPAC Name N-(4-methyl-1,3-thiazol-2-yl)-2-(6-phenylpyridazin-3-yl)sulfanylacetamide
SMILES Cc1csc(NC(=O)CSc2ccc(nn2)c3ccccc3)n1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H14N4OS2/c1-11-9-23-16(17-11)18-14(21)10-22-15-8-7-13(19-20-15)12-5-3-2-4-6-12/h2-9H,10H2,1H3,(H,17,18,21)
Formula Weight 342.0609
Chemical Name or Material N-(4-Methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]-acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.