missing translation for 'onlineSavingsMsg'
Learn More

N-[4-Methyl-3-(4-pyridin-3-yl-pyrimidin-2-ylamino)-phenyl]-4-chloromethyl Benzamide, TRC

Product Code. 30477649
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477649 5 mg 5mg
30474353 10 mg 10mg
30481626 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477649 Supplier Toronto Research Chemicals Supplier No. TRCM3259435MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 404844-11-7
Molecular Formula C24 H20 Cl N5 O
Molecular Weight (g/mol) 429.9
Synonym Benzamide, 4-(chloromethyl)-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]-, 2-Methyl-5-[[4-(chloromethyl)benzoyl]amino]-N-[4-(pyridin-3-yl)pyrimidin-2-yl]benzenamine, 4-(Chloromethyl)-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide, 4-Chloromethyl-N-[4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]phenyl]benzamide, N-[5-[[4-(Chloromethyl)benzoyl]amino]-2-methylphenyl]-4-(3-pyridyl)-2-pyrimidineamine
IUPAC Name 4-(chloromethyl)-N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide
SMILES Cc1ccc(NC(=O)c2ccc(CCl)cc2)cc1Nc3nccc(n3)c4cccnc4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C24H20ClN5O/c1-16-4-9-20(28-23(31)18-7-5-17(14-25)6-8-18)13-22(16)30-24-27-12-10-21(29-24)19-3-2-11-26-15-19/h2-13,15H,14H2,1H3,(H,28,31)(H,27,29,30)
Formula Weight 429.1356
Chemical Name or Material 4-(Chloromethyl)-N-[4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]phenyl]benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.