missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Pentynyl)phthalimide, 97%

Product Code. 15460238
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460238 25 g 25g
15450238 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15460238 Supplier Thermo Scientific Alfa Aesar Supplier No. H53432.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6097-07-0 | C13H11NO2 | 213.236 g/mol

N-(4-Pentynyl)phthalimide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(4-Pentynyl)phthalimide is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6097-07-0
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.236
MDL Number MFCD06798110
InChI Key YNZIPXLLPFYDGM-UHFFFAOYSA-N
Synonym n-4-pentynyl phthalimide, 2-pent-4-ynyl isoindoline-1,3-dione, 1-phthalimido-4-pentyne, 2-pent-4-yn-1-yl isoindoline-1,3-dione, n-4-pentynyl phthalimide 97, 2-pent-4-yn-1-yl isoindole-1,3-dione, 5-phthalimido-1-pentyne, 2-pent-4-ynyl-isoindole-1,3-dione
PubChem CID 11769822
IUPAC Name 2-pent-4-ynylisoindole-1,3-dione
SMILES C#CCCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 87°C to 91°C
Quantity 25 g
Formula Weight 213.24
Percent Purity 97%
Chemical Name or Material N-(4-Pentynyl)phthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.