missing translation for 'onlineSavingsMsg'
Learn More

N-{[5-(2-Furyl)thien-2-yl]methyl}-N-methylamine, 97%, Thermo Scientific™

Product Code. 10642325
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10642325 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10642325 Supplier Thermo Scientific Maybridge Supplier No. CC47746DA
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 869901-16-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H11NOS
MDL Number MFCD08690272
Quantity 1 g
Synonym n-5-2-furyl thien-2-yl methyl-n-methylamine, 5-furan-2-yl thiophen-2-yl methyl methyl amine, 5-2-furyl 2-thienyl methyl methylamine, 2-thiophenemethanamine,5-2-furanyl-n-methyl, 1-5-furan-2-yl thiophen-2-yl-n-methylmethanamine
InChI Key REBAUXOPYFOJIE-UHFFFAOYSA-N
SMILES CNCC1=CC=C(S1)C1=CC=CO1
IUPAC Name 1-[5-(furan-2-yl)thiophen-2-yl]-N-methylmethanamine
Molecular Weight (g/mol) 193.26
PubChem CID 18525822
Formula Weight 193.27
Percent Purity 97%
Chemical Name or Material N-{[5-(2-furyl)thien-2-yl]methyl}-N-methylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.