missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(5-Chloro-2-methylphenyl)-2-fluorobenzamide, 97%

Brand:  Alfa Aesar™ H59193.03

Code : NEW

Additional Details : CAS Number : 331436-17-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415438

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(5-Chloro-2-methylphenyl)-2-fluorobenzamide
331436-17-0
263.7
MFCD00684374

Specifications

N-(5-Chloro-2-methylphenyl)-2-fluorobenzamide
Solid
263.7
1g
331436-17-0
97%
MFCD00684374
Videos
SDS