missing translation for 'onlineSavingsMsg'
Learn More

N-(5-chloro-2-pyridyl)bis(trifluoromethanesulfonimide)

Product Code. 10475095 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10475095 5 g Glass bottle 5g
10085113 1 g Glass bottle 1g
11936331 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10475095 Supplier Thermo Scientific Acros Supplier No. 378800050
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 145100-51-2 | C7H3ClF6N2O4S2 | 392.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145100-51-2
Molecular Formula C7H3ClF6N2O4S2
Molecular Weight (g/mol) 392.69
MDL Number MFCD00191833
InChI Key TUFGVZMNGTYAQD-UHFFFAOYSA-N
Synonym 2-n,n-bis trifluoromethylsulphonyl amino-5-chloropyridine, 2-n,n-bis trifluoromethanesulfonyl amino-5-chloropyridine, comins' reagent, n-5-chloro-2-pyridyl triflimide, 2-n,n-bis trifluoromethylsulfonyl amino-5-chloropyridine, comins triflating reagent, n-5-chloro-2-pyridyl bis trifluoromethanesulfonimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, n-5-chloropyridin-2-yl triflimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide
PubChem CID 388544
IUPAC Name N-(5-chloropyridin-2-yl)-1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C1=CC(=NC=C1Cl)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

Specifications

Melting Point 45.0°C to 47.0°C
Boiling Point 77.0°C to 90.0°C (0.2 mmHg)
Packaging Glass bottle
Quantity 5 g
Formula Weight 392.69
Chemical Name or Material N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonimide)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.