missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Hexynyl)phthalimide, 97%

Product Code. 15430368
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430368 5 g 5g
15440368 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430368 Supplier Thermo Scientific Alfa Aesar Supplier No. H53489.06
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6097-08-1 | C14H13NO2 | 227.26 g/mol

6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6097-08-1
MDL Number MFCD00671372

Specifications

Melting Point 68°C to 72°C
Molecular Formula C10H10O3
Quantity 5 g
Synonym 3-methoxycinnamic acid, 3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl prop-2-enoic acid, trans-3-methoxycinnamic acid, m-methoxycinnamic acid, trans-3-3-methoxyphenyl acrylic acid, e-3-3-methoxyphenyl acrylic acid, 2-propenoic acid, 3-3-methoxyphenyl-, 2e, trans-m-methoxycinnamic acid
InChI Key LZPNXAULYJPXEH-AATRIKPKSA-N
SMILES COC1=CC=CC(=C1)C=CC(=O)O
IUPAC Name (E)-3-(3-methoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 178.19
PubChem CID 637668
Formula Weight 227.26
Percent Purity 97%
Chemical Name or Material N-(5-Hexynyl)phthalimide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.