missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-(2R)-bornane-10,2-sultam, Thermo Scientific™

Product Code. 10225541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10225541 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10225541 Supplier Thermo Scientific Acros Supplier No. 337760010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 141993-16-0
Packaging Glass bottle
Molecular Formula C12H19NO3S
MDL Number MFCD00269649,MFCD00269650,MFCD00269649
Quantity 1 g
Synonym n-acetyl-2r-bornane-10,2-sultam
InChI Key FECARJHVTMSYSJ-UMNHJUIQSA-N
SMILES CC(=O)N1[C@@H]2C[C@@H]3CC[C@@]2(CS1(=O)=O)C3(C)C
IUPAC Name (1R,5R,7S)-4-acetyl-10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-3,3-dione
Molecular Weight (g/mol) 257.35
PubChem CID 60161326
Formula Weight 257.35
Chemical Name or Material N-Acetyl-(2R)-bornane-10, 2-sultam
Show More Show Less
compliance-icons
Product Identifier
  • N-Acetyl-(2R)-bornane-10,2-sultam
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.