missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-4-benzoquinone Imine, TRC

Product Code. 17973375
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973375 5 mg 5mg
17942345 25 mg 25mg
17963465 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973375 Supplier Toronto Research Chemicals Supplier No. TRCA1700005MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50700-49-7
Molecular Formula C8 H7 N O2
Molecular Weight (g/mol) 149.15
Synonym Acetamide, N-(4-oxo-2,5-cyclohexadien-1-ylidene)-, N-(4-Oxo-2,5-cyclohexadien-1-ylidene)acetamide, Acetimidoquinone, N-Acetyl-1,4-benzoquinonimine, N-Acetyl-p-benzoquinonimine, N-(4-oxocyclohexa-2,5-dien-1-ylidene)acetamide, N-Acetyl-p-benzoquinone Imine
IUPAC Name N-(4-oxocyclohexa-2,5-dien-1-ylidene)acetamide
SMILES CC(=O)N=C1C=CC(=O)C=C1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C8H7NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5H,1H3
Formula Weight 149.0477
Chemical Name or Material N-Acetyl-4-benzoquinone Imine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.