missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-4-piperidone, 97%

Product Code. 10173890 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10173890

Brand: Thermo Scientific Acros 166821000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32161-06-1 | C7H11NO2 | 141.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32161-06-1
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00006190
InChI Key NNFOVLFUGLWWCL-UHFFFAOYSA-N
Synonym 1-acetyl-4-piperidone, n-acetyl-4-piperidone, 4-piperidinone, 1-acetyl, 1-acetyl-4-piperidinone, 1-acetyl-piperidin-4-one, 1-acetyl-4-oxopiperidine, n-acetyl-4-piperidinone, n-acetyl4-piperidone, pubchem19615, 1-acetylpiperid-4-one
PubChem CID 122563
IUPAC Name 1-acetylpiperidin-4-one
SMILES CC(=O)N1CCC(=O)CC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Orange
Density 1.1400g/mL
Boiling Point 218°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5015 to 1.5035
Quantity 100 g
Specific Gravity 1.14
Formula Weight 141.17
Percent Purity 97%
Physical Form Oily Liquid
Chemical Name or Material N-Acetyl-4-piperidone, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.