missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-Arg-Glu-Lys-Arg-7-amino-4-(trifluoromethyl)coumarin

Product Code. 15400347
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400347 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15400347 Supplier Thermo Scientific Chemicals Supplier No. J64106.MA
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

C35H51F3N12O9

N-Acetyl-Arg-Glu-Lys-Arg-7-amino-4-(trifluoromethyl)coumarin is employed as a fluorogenic cathepsin D substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A fluorogenic substrate suitable for the assay of furin
Molecular Formula C35H51F3N12O9
Formula Weight 840.80g/mol
Synonym Ac-REKR-AFC; Ac-Arg-Glu-Lys-Arg-AFC
Sensitivity Light Sensitive
Solubility Soluble in DMSO or DMF
Product Type Signal Transduction Reagent
Form Solid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.