missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl D-Alpha-Methyl DOPA Dimethyl Ether, TRC

Product Code. 30315893
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315893 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315893 Supplier Toronto Research Chemicals Supplier No. TRCA18666510MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl D-alpha-Methyl DOPA Dimethyl Ether
CAS 2503-39-1
Molecular Formula C14H19NO5
Quantity 10 mg
Synonym N-Acetyl-3-methoxy-O,alpha-dimethyl-D-tyrosine, N-Acetyl-3-(3,4-dimethoxyphenyl)-2-methyl-alanine
InChI Formula InChI=1S/C14H19NO5/c1-9(16)15-14(2,13(17)18)8-10-5-6-11(19-3)12(7-10)20-4/h5-7H,8H2,1-4H3,(H,15,16)(H,17,18)/t14-/m1/s1
SMILES COc1ccc(C[C@@](C)(NC(=O)C)C(=O)O)cc1OC
IUPAC Name (2R)-2-acetamido-3-(3,4-dimethoxyphenyl)-2-methylpropanoic acid
Molecular Weight (g/mol) 281.3
Formula Weight 281.1263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.