missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-D-glucosamine, 98+%

Product Code. 11438770
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438770 100 g 100g
11479983 10 g 10g
11489983 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438770 Supplier Thermo Scientific Chemicals Supplier No. A13047.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-Acetyl-D-glucosamine, CAS # 72-87-7, is the D isomer of the glucose-derived monomer found in structural polysaccharides.

N-Acetyl-D-glucosamine is used in the identification and characterization of N-acetyl-beta-D-hexoaminidase. It is also used in the treatment of osteoarthritis and inflammatory bowel disease (IBD), including ulcerative colitis and Crohn's disease. Further, it is used as a substrate in sialic acid production, cosmetics and in drug development research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• N-Acetyl-D-glucosamine is the acetylated form of glucosamine, a glucose derivative

• It has a role as a prokaryotic metabolism and monomer used in chitin, hyaluronic acid, and polymers within the cell wall

Application

• N-Acetyl-D-Glucosamine can be used for molecular binding studies; research has shown that it attaches to certain microbes and their pathogens

• It is useful for biochemical characterization studies of related glucosamines

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00061615
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Melting Point 213°C (decomposition)
Packaging Glass bottle
Quantity 100 g
Beilstein 1727589
Sensitivity Hygroscopic
Solubility Information Soluble in concentrated hydrochloric acid,sulfuric acid,phosphoric acid and formic acid. Insoluble in water,dilute acids,dilute alkalis,concentrated alkalis and organic solvents.
Formula Weight 221.21
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material N-Acetyl-D-glucosamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.