missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-D-glucosamine, 98+%

Product Code. 11479983
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
This item is not returnable. View return policy

Product Code. 11479983

Brand: Thermo Scientific Chemicals A13047.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

N-Acetyl-D-glucosamine, CAS # 72-87-7, is the D isomer of the glucose-derived monomer found in structural polysaccharides.

N-Acetyl-D-glucosamine is used in the identification and characterization of N-acetyl-beta-D-hexoaminidase. It is also used in the treatment of osteoarthritis and inflammatory bowel disease (IBD), including ulcerative colitis and Crohn's disease. Further, it is used as a substrate in sialic acid production, cosmetics and in drug development research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• N-Acetyl-D-glucosamine is the acetylated form of glucosamine, a glucose derivative

• It has a role as a prokaryotic metabolism and monomer used in chitin, hyaluronic acid, and polymers within the cell wall

Application

• N-Acetyl-D-Glucosamine can be used for molecular binding studies; research has shown that it attaches to certain microbes and their pathogens

• It is useful for biochemical characterization studies of related glucosamines

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00061615
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

Especificaciones

Melting Point 213°C (decomposition)
Assay Percent Range ≥98%
Quantity 10 g
Beilstein 1727589
Sensitivity Hygroscopic
Solubility Information Soluble in concentrated hydrochloric acid,sulfuric acid,phosphoric acid and formic acid. Insoluble in water,dilute acids,dilute alkalis,concentrated alkalis and organic solvents.
Formula Weight 221.21
Chemical Name or Material N-Acetyl-D-glucosamine

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado