missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-mannosamine monohydrate, 99%

Product Code. 11367606
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367606 5 g 5g
11377606 250 mg 250mg
11357606 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367606 Supplier Thermo Scientific Alfa Aesar Supplier No. L11167.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1071625-31-4 | C8H17NO7 | 239.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071625-31-4
Molecular Formula C8H17NO7
Molecular Weight (g/mol) 239.22
MDL Number MFCD00149493,MFCD00046758,MFCD00149493,MFCD00069808
InChI Key VVQPUTSNIMAJPT-UHFFFAOYNA-N
Synonym n-acetylmannosamine
PubChem CID 11908605
IUPAC Name N-[(2R,3S,4S,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES O.CC(=O)NC1C(O)OC(CO)C(O)C1O

Specifications

Melting Point ∼130°C (decomposition)
Packaging Glass bottle
Quantity 5 g
Beilstein 1346524
Formula Weight 239.24 (221.22 anhydrous)
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-Acetyl-D-mannosamine monohydrate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.