missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-D-sphingosine, 98%

Product Code. 15453289
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453289 10 mg 10mg
15443289 1 mg 1mg
15433289 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15453289 Supplier Thermo Scientific Chemicals Supplier No. J60534.MA
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3102-57-6 | C20H39NO3, C20H39NO3 | 341.54 g/mol

Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells
  • Activates protein phosphatases
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3102-57-6
Molecular Formula C20H39NO3
Synonym Acetyl ceramide; C2-Ceramide

Specifications

Quantity 10 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases
Formula Weight 341.53g/mol
Melting Point 77°C to 78°C
Solubility Soluble in ethanol, methanol, chloroform or DMSO at approximately 10mg/ml
Product Type Signal Transduction Reagent
Form Powder
Assay 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.