missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-d3-5-hydroxytryptamine, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-d3-5-hydroxytryptamine |
| CAS | 2001098-07-1 |
| Molecular Formula | C12 D3 H11 N2 O2 |
| Quantity | 0.005 g |
| Synonym | N-[2-(5-Hydroxy-1 H-indol-3-yl)ethyl]acetamide-d3, 5-Hydroxy-N-acetyltryptamine-d3, 5-Hydroxymelatonin-d3, N-Acetylserotonin-d3, N-[2-(5-Hydroxy-1 H-indol-3-yl)ethyl]trideuterioacetamide, 5-Hydroxymelatonin-d3, N-(Acetyl-d3)-2-(5-hydroxyindol-3-yl)ethylamine, N-(Acetyl-d3)-5-hydroxytryptamine, N-Acetyl-5-hydroxytryptamine-D3 (N-acetyl-D3), N-Acetyl-d3-serotonin, Normelatonin-d3, O-Demethylmelatonin-d3 |
| InChI Formula | InChI=1 S/C12H14N2O2/c1-8(15)13-5-4-9-7-14-12-3-2-10(16)6-11(9)12/h2-3,6-7,14,16 H,4-5H2,1H3,(H,13,15)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)NCCc1c[nH]c2ccc(O)cc12 |
| IUPAC Name | 2,2,2-trideuterio-N-[2-(5-hydroxy-1 H-indol-3-yl)ethyl]acetamide |
| Molecular Weight (g/mol) | 221.27 |
| Formula Weight | 221.1244 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction