missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-DL-alanine, 97+%

Product Code. 11348375
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348375 25 g 25g
11338375 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348375 Supplier Thermo Scientific Chemicals Supplier No. L10329.14
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1115-69-1 | C5H9NO3 | 131.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-69-1
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.131
MDL Number MFCD00037238
InChI Key KTHDTJVBEPMMGL-UHFFFAOYSA-N
Synonym n-acetyl-dl-alanine, ac-dl-ala-oh, 2-acetylaminopropionic acid, n-acetylalanine, acetyl-dl-alanine, dl-alanine, n-acetyl, 2-acetylamino-propionic acid, n-acetyl-s-alanine, n-alpha-acetyl-dl-alanine, n-acetyl-dl-2-aminopropionic acid
PubChem CID 7345
IUPAC Name 2-acetamidopropanoic acid
SMILES CC(C(=O)O)NC(=O)C

Specifications

Melting Point 135°C to 139°C
Beilstein 774333
Quantity 25 g
Formula Weight 131.13
Percent Purity ≥97%
Chemical Name or Material N-Acetyl-DL-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.