missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-methionine, TRC

Product Code. 30353015
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353015 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353015 Supplier Toronto Research Chemicals Supplier No. TRCC3826082.5G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl-DL-methionine
CAS 1115-47-5
Purity Grade Notes HPLC
Molecular Formula C7 H13 N O3 S
Quantity 2.5 g
Synonym N-Acetylmethionine, DL-Methionine, N-acetyl- (6CI), Methionine, N-acetyl-, DL- (8CI), Acetyl-DL-methionine, DL-N-acetylmethionine, N-Acetyl-DL-methionine, NSC 7633
InChI Formula InChI=1S/C7H13NO3S/c1-5(9)8-6(7(10)11)3-4-12-2/h6H,3-4H2,1-2H3,(H,8,9)(H,10,11)
SMILES CSCCC(NC(=O)C)C(=O)O
IUPAC Name 2-acetamido-4-methylsulfanylbutanoic acid
Molecular Weight (g/mol) 191.25
Formula Weight 191.0616
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.