missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-DL-tryptophan, 98+%, extra pure

Product Code. 10752461 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10752461 25 g 25g
10013040 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10752461 Supplier Thermo Scientific Chemicals Supplier No. 227150250
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 87-32-1 | C13H14N2O3 | 246.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-32-1
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00005644
InChI Key DZTHIGRZJZPRDV-UHFFFAOYSA-N
Synonym n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
PubChem CID 2002
ChEBI CHEBI:70976
IUPAC Name 2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 204.0°C to 206.0°C
Color White to Yellow
Loss on Drying 0.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: 0,51g/L (20°C). Other solubilities: soluble in methanol
Formula Weight 246.27
Percent Purity 98+%
Grade Extra Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Acetyl-DL-tryptophan, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.