missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-tryptophan, 98+%, extra pure, Thermo Scientific™

Product Code. 15295978 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15295978 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15295978 Supplier Thermo Scientific Acros Supplier No. 227150010
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 87-32-1
Melting Point 204.0°C to 206.0°C
Loss on Drying 0.5% max. (105°C)
Infrared Spectrum Authentic
Molecular Formula C13H14N2O3
MDL Number MFCD00005644
Quantity 1 kg
Synonym n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
Solubility Information Solubility in water: 0,51g/L (20°C). Other solubilities: soluble in methanol
InChI Key DZTHIGRZJZPRDV-UHFFFAOYSA-N
SMILES CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
IUPAC Name 2-acetamido-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 246.27
PubChem CID 2002
ChEBI CHEBI:70976
Formula Weight 246.27
Percent Purity 98+%
Grade Extra Pure
Chemical Name or Material N-Acetyl-DL-tryptophan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.