missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-tryptophan, 99%

Product Code. 11488187
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488187 100 g 100g
11478187 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488187 Supplier Thermo Scientific Alfa Aesar Supplier No. A17562.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-32-1 | C13H14N2O3 | 246.27 g/mol

N-Acetyl-DL-tryptophan is produced via chemical synthesis using the standard amino acid, tryptophan. It can be used in downstream applications of biopharmaceutical production as a transfer agent or stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-DL-tryptophan is produced via chemical synthesis using the standard amino acid, tryptophan. It can be used in downstream applications of biopharmaceutical production as a transfer agent or stabilizer.

Solubility
Soluble in 1N Sodium Hydroxide, water, and methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-32-1
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00005644
InChI Key DZTHIGRZJZPRDV-UHFFFAOYSA-N
Synonym n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
PubChem CID 2002
ChEBI CHEBI:70976
IUPAC Name 2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point ∼205°C (decomposition)
Beilstein 89478
Solubility Information Soluble in 1N Sodium Hydroxide,water,and methanol.
Quantity 100 g
Formula Weight 246.27
Percent Purity 99%
Chemical Name or Material N-Acetyl-DL-tryptophan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.