missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-glutamic acid, 99%

Product Code. 11455657
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455657 100 g 100g
11445657 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455657 Supplier Thermo Scientific Chemicals Supplier No. B23621.22
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1188-37-0 | C7H11NO5 | 189.167 g/mol

N-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.

Solubility
Soluble in water. 2.7 g/100 mL (20°C)

Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1188-37-0
Molecular Formula C7H11NO5
Molecular Weight (g/mol) 189.167
MDL Number MFCD00002802
InChI Key RFMMMVDNIPUKGG-YFKPBYRVSA-N
Synonym n-acetyl-l-glutamic acid, acetylglutamic acid, n-acetyl-l-glutamate, n-acetylglutamic acid, n-acetylglutamate, acetyl glutamic acid, acetyl-l-glutamic acid, l-glutamic acid, n-acetyl, l-n-acetylglutamic acid, ac-glu-oh
PubChem CID 70914
ChEBI CHEBI:17533
IUPAC Name (2S)-2-acetamidopentanedioic acid
SMILES CC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 194°C to 199°C
Beilstein 1727473
Solubility Information Soluble in water. 2.7g/100 mL (20°C)
Quantity 100 g
Formula Weight 189.17
Optical Rotation −16.5° (c=1 in Water)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-glutamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.