missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-glutamine, 97%, Thermo Scientific™

Product Code. 15234884 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
Product Code. Quantity Packaging unitSize
15234884 5 g Glass bottle 5g
15244884 25 g Glass bottle 25g
15264884 100 g Glass bottle 100g
3 options
This item is not returnable. View return policy

Product Code. 15234884

Brand: Thermo Scientific Acros 451640050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 35305-74-9
Molecular Formula C7H12N2O4
Molecular Weight (g/mol) 188.18
MDL Number MFCD00038159
InChI Key KSMRODHGGIIXDV-YFKPBYRVSA-N
Synonym n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine
PubChem CID 182230
ChEBI CHEBI:21553
SMILES CC(=O)N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 206°C
Ignition Residue 0.2% max
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
IUPAC Name (2S)-4-carbamoyl-2-acetamidobutanoic acid
Formula Weight 188.18
Percent Purity 97%
Chemical Name or Material N-Acetyl-L-glutamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.