missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-leucine, 99%

Product Code. 10377181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10377181 25 g Glass bottle 25g
10235961 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10377181 Supplier Thermo Scientific Chemicals Supplier No. 347010250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1188-21-2 | C8H15NO3 | 173.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1188-21-2
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00065131
InChI Key WXNXCEHXYPACJF-JLDDOWRYNA-N
Synonym n-acetyl-l-leucine, acetyl-l-leucine, ac-leu-oh, n-acetylleucine, s-2-acetamido-4-methylpentanoic acid, n-acetyl-leu, 2s-2-acetamido-4-methylpentanoic acid, n-acetyl-leucine, leucine, n-acetyl-, l, n-acetyl-l-leucin
PubChem CID 70912
ChEBI CHEBI:17786
IUPAC Name (2S)-2-acetamido-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC(C)=O)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 190.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH(NHCOCH3)COOH
Quantity 25 g
Specific Rotation − 24.50
Solubility Information (5% in ethanol) Clear colorless
Formula Weight 173.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-leucine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.