missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-leucine, 99%

Product Code. p-7046314
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378825 5 g 5g
11388825 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378825 Supplier Thermo Scientific Chemicals Supplier No. L13926.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1188-21-2
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00065131
InChI Key WXNXCEHXYPACJF-JLDDOWRYNA-N
Synonym n-acetyl-l-leucine, acetyl-l-leucine, ac-leu-oh, n-acetylleucine, s-2-acetamido-4-methylpentanoic acid, n-acetyl-leu, 2s-2-acetamido-4-methylpentanoic acid, n-acetyl-leucine, leucine, n-acetyl-, l, n-acetyl-l-leucin
PubChem CID 70912
ChEBI CHEBI:17786
IUPAC Name (2S)-2-acetamido-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC(C)=O)C(O)=O

Specifications

Melting Point 177°C to 179°C
Beilstein 1724849
Quantity 5 g
Formula Weight 173.21
Optical Rotation −23° (c=5 in Ethanol)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-leucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.