missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-methionine, 99%

Product Code. 11445737
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445737 5 g 5g
11455737 25 g 25g
11465737 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445737 Supplier Thermo Scientific Alfa Aesar Supplier No. B24312.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65-82-7 | C7H13NO3S | 191.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-82-7
Molecular Formula C7H13NO3S
Molecular Weight (g/mol) 191.25
MDL Number MFCD00064441
InChI Key XUYPXLNMDZIRQH-LURJTMIESA-N
Synonym n-acetyl-l-methionine, n-acetylmethionine, acetyl-l-methionine, ac-met-oh, l-n-acetyl methionine, acetylmethionine, methionamine, methionine, n-acetyl-, l, thiomedon, l-methionine, n-acetyl
PubChem CID 448580
ChEBI CHEBI:21557
IUPAC Name (2S)-2-acetamido-4-methylsulfanylbutanoic acid
SMILES CSCC[C@H](NC(C)=O)C(O)=O

Specifications

Melting Point 103°C to 107°C
Beilstein 1725552
Merck Index 14,96
Quantity 5 g
Formula Weight 191.25
Optical Rotation −21° (c=1.3 in Water)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-methionine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.