missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-phenylalanyl-3,5-diiodo-L-tyrosine, 95%, Thermo Scientific™

Product Code. 10701321 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
250mg; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10701321 250mg; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10701321 Supplier Thermo Scientific Acros Supplier No. 227652500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3786-08-1
Color White
Quantity 250 mg
Molecular Weight (g/mol) 622.2
Percent Purity 95%
Physical Form Crystals or Crystalline Powder
Chemical Name or Material N-Acetyl-L-phenylalanyl-3, 5-diiodo-L-tyrosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.