missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-l-proline, 99%

Product Code. 10578093 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10578093 1 g Glass bottle 1g
10005582 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10578093 Supplier Thermo Scientific Acros Supplier No. 376640010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68-95-1 | C7H11NO3 | 157.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-95-1
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00020837
InChI Key GNMSLDIYJOSUSW-UHFFFAOYNA-N
Synonym n-acetyl-l-proline, acetylproline, ac-pro-oh, n-acetylproline, 1-acetyl-l-proline, acetyl proline, l-proline, 1-acetyl, acetyl-l-proline, 2s-1-acetylpyrrolidine-2-carboxylic acid, s-1-acetylpyrrolidine-2-carboxylic acid
PubChem CID 66141
ChEBI CHEBI:21560
IUPAC Name (2S)-1-acetylpyrrolidine-2-carboxylic acid
SMILES CC(=O)N1CCCC1C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 115°C to 117°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition −86° (20°C c=1 EtOH)
Specific Rotation -86
Formula Weight 157.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-Acetyl-L-proline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.