missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tryptophan methyl ester, 95%

Product Code. 15492839
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492839 25 g 25g
15482839 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492839 Supplier Thermo Scientific Alfa Aesar Supplier No. H66937.14
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2824-57-9 | C14H16N2O3 | 260.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2824-57-9
Molecular Formula C14H16N2O3
Molecular Weight (g/mol) 260.29
MDL Number MFCD00022759
InChI Key XZECNVJPYDPBAM-ZDUSSCGKSA-N
Synonym methyl n-acetyl-l-tryptophanate, ac-trp-ome, methyl acetyltryptophanate, l-tryptophan, n-acetyl-, methyl ester, unii-mup4y18zq5, n-acetyltryptophan methyl ester, n-acetyl-l-tryptophan methyl ester, mup4y18zq5, methyl 2s-2-acetamido-3-1h-indol-3-yl propanoate, l-tryptophan,n-acetyl-, methyl ester
PubChem CID 102337
IUPAC Name methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
SMILES COC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(C)=O

Specifications

Color White
Quantity 25 g
Formula Weight 260.29
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Acetyl-L-tryptophan methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.