missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-S-phenyl-d5-DL-cysteine, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-S-phenyl-d5-DL-cysteine |
| Molecular Formula | C11 D5 H8 N O3 S |
| Quantity | 0.01 g |
| Synonym | N-Acetyl-S-(phenyl-d5)-DL-cysteine, N-Acetyl-S-phenyl-DL-cysteine-d5, N-Acetyl-S-phenyl-DL-cysteine-D5 (phenyl-D5), 2-Acetamido-3-(phenyl-d5-sulfanyl)propanoic acid, (RS)-S-Phenyl-d5-mercapturic acid |
| InChI Formula | InChI=1 S/C11H13NO3S/c1-8(13)12-10(11(14)15)7-16-9-5-3-2-4-6-9/h2-6,10 H,7H2,1H3,(H,12,13)(H,14,15)/i2D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(SCC(NC(=O)C)C(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | 2-acetamido-3-(2,3,4,5,6-pentadeuteriophenyl)sulfanylpropanoic acid |
| Molecular Weight (g/mol) | 244.322 |
| Formula Weight | 244.093 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction