missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-Ser-Asp-Lys-Pro

Product Code. 15437789
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 15437789

Brand: Thermo Scientific Chemicals J66430.#0

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

N-Acetyl-Ser-Asp-Lys-Pro inhibits the entry of pluripotent hemopoietic stem cells into S-phase of the cell cycle and protects against Ara-C lethality in mice. It is a specific substrate for the N-terminal active site of angiotensin converting enzyme, which is responsible for its degradation in vivo.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-Ser-Asp-Lys-Pro inhibits the entry of pluripotent hemopoietic stem cells into S-phase of the cell cycle and protects against Ara-C lethality in mice. It is a specific substrate for the N-terminal active site of angiotensin
TRUSTED_SUSTAINABILITY

Specifications

CAS 127103-11-1
Color White
Content And Storage Store at -20°C
Format Solid
Molecular Weight (g/mol) 487.51
Product Type N-Acetyl-Ser-Asp-Lys-Pro
Quantity 5 mg

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.