missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylcaprolactam, 99%

Product Code. 11473797
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473797 25 g 25g
11483797 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473797 Supplier Thermo Scientific Alfa Aesar Supplier No. A18257.14
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1888-91-1 | C8H13NO2 | 155.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1888-91-1
Molecular Formula C8H13NO2
Molecular Weight (g/mol) 155.20
MDL Number MFCD00003262
InChI Key QISSLHPKTCLLDL-UHFFFAOYSA-N
Synonym n-acetylcaprolactam, acetylcaprolactam, n-acetyl-6-caprolactam, 2h-azepin-2-one, 1-acetylhexahydro, acetylkaprolaktam, n-acetylhexanelactam, acetylkaprolaktam czech, 1-acetylhexahydro-2h-azepin-2-one, n-acetyl-epsilon-caprolactam, n-acetyl-.epsilon.-caprolactam
PubChem CID 15904
IUPAC Name 1-acetylazepan-2-one
SMILES CC(=O)N1CCCCCC1=O

Specifications

Density 1.094
Boiling Point 124°C (12 mmHg)
Flash Point 122°C (252°F)
Refractive Index 1.489
Quantity 25 g
Beilstein 119598
Formula Weight 155.2
Percent Purity 99%
Chemical Name or Material N-Acetylcaprolactam
compliance-icons compliance-icons
Product Identifier
  • N-Acetylcaprolactam
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N-Acetylcaprolactam

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.