missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylethylenediamine, tech. 90%

Product Code. 11373027
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373027 25 g 25g
11363027 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373027 Supplier Thermo Scientific Alfa Aesar Supplier No. L13398.14
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1001-53-2 | C4H10N2O | 102.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1001-53-2
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.137
MDL Number MFCD00008163
InChI Key DAKZISABEDGGSV-UHFFFAOYSA-N
Synonym n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine
PubChem CID 66082
IUPAC Name N-(2-aminoethyl)acetamide
SMILES CC(=O)NCCN

Specifications

Melting Point 44°C to 50°C
Boiling Point 128°C (3 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3CONHCH2CH2NH2
Quantity 25 g
UN Number UN3259
Beilstein 1743120
Sensitivity Hygroscopic
Formula Weight 102.14
Percent Purity 90%
Grade Technical
Chemical Name or Material N-Acetylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.