missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylneuraminic acid, 99%, Thermo Scientific Chemicals

Product Code. 10276520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10276520 25 mg Glass bottle 25mg
10397980 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10276520 Supplier Thermo Scientific Acros Supplier No. 227040250
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-48-6
Molecular Formula C11H19NO9
Molecular Weight (g/mol) 309.271
MDL Number MFCD00006620
InChI Key SQVRNKJHWKZAKO-LFIUDZTESA-N
Synonym n-acetylneuramic acid, spectrum_000450, spectrum2_000559, spectrum4_000432, spectrum5_001326, spectrum2300147, 2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
PubChem CID 126963458
IUPAC Name (4S,5S,6R)-5-acetamido-2,4-dihydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
SMILES CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 mg
Merck Index 15, 8623
Specific Rotation -30° to -34° (20°C, 589nm) (c=2, water)
Formula Weight 309.26
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material N-Acetylneuraminic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.